accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6972-69-6

IUPAC Name: N,N-dimethylpyridine-3-carboxamide
Synonyms: N,N-dimethylnicotinamide
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: FJEVKYZLIRAAKE-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=CC=CN=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results