Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylnicotinamide 99.0+%, TCI America™
SDP

Catalog No. D075810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D075810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D075810G Supplier TCI America Supplier No. D075810G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6972-69-6
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00059169
InChI Key FJEVKYZLIRAAKE-UHFFFAOYSA-N
PubChem CID 81444
IUPAC Name N,N-dimethylpyridine-3-carboxamide
SMILES CN(C)C(=O)C1=CN=CC=C1

Specifications

Melting Point 44°C
Color White-Yellow
Boiling Point 147°C
Quantity 10 g
Formula Weight 150.18
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dimethylnicotinamide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.