accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3COCH3H2C

CAS RN 6976-91-6

IUPAC Name: N,N,2-trimethylprop-2-enamide
Synonyms: N,N,2-trimethylprop-2-enamide
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: QRWZCJXEAOZAAW-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results