Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylmethacrylamide 97.0+%, TCI America™
SDP

Catalog No. D074510ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D074510ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D074510ML Supplier TCI America Supplier No. D074510ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6976-91-6
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00059328
InChI Key QRWZCJXEAOZAAW-UHFFFAOYSA-N
PubChem CID 81465
IUPAC Name N,N,2-trimethylprop-2-enamide
SMILES CC(=C)C(=O)N(C)C

Specifications

Color Yellow
Boiling Point 67°C
Quantity 10 mL
Formula Weight 113.16
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylmethacrylamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.