accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3ONCH3OHNO

CAS RN 6979-97-1

IUPAC Name: [3-(acetyloxy)-5-(5-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)oxolan-2-yl]methyl acetate
Synonyms: [3-(acetyloxy)-5-(5-methyl-2,4-dioxo-3H-pyrimidin-1-yl)oxolan-2-yl]methyl acetate
Molecular Weight (g/mol): 326.31
Molecular Formula: C14H18N2O7
InChi Key: RGVBNBFNSBMXID-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(CC1OC(C)=O)N1C=C(C)C(=O)NC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)