accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 698-10-2

IUPAC Name: 5-ethyl-3-hydroxy-4-methyl-2,5-dihydrofuran-2-one
Synonyms: 5-ethyl-3-hydroxy-4-methyl-5H-furan-2-one
Molecular Weight (g/mol): 142.15
Molecular Formula: C7H10O3
InChi Key: IUFQZPBIRYFPFD-UHFFFAOYNA-N
SMILES: CCC1OC(=O)C(O)=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results