accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFOFFFOFFFFNFFF

CAS RN 69804-19-9

IUPAC Name: 2,2,3,3-tetrafluoro-3-({1,1,1,2,3,3-hexafluoro-3-[(1,2,2-trifluoroethenyl)oxy]propan-2-yl}oxy)propanenitrile
Synonyms: 2,2,3,3-tetrafluoro-3-({1,1,1,2,3,3-hexafluoro-3-[(1,2,2-trifluoroethenyl)oxy]propan-2-yl}oxy)propanenitrile
Molecular Weight (g/mol): 389.07
Molecular Formula: C8F13NO2
InChi Key: LYIPDZSLYLDLCU-UHFFFAOYNA-N
SMILES: FC(F)=C(F)OC(F)(F)C(F)(OC(F)(F)C(F)(F)C#N)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results