Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3-Tetrafluoro-3-[[1,1,1,2,3,3-hexafluoro-3-[(1,2,2-trifluorovinyl)oxy]propan-2-yl]oxy]propionitrile 93.0+%, TCI America™
SDP

Catalog No. T34935G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34935G 5 g
T349325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34935G Supplier TCI America Supplier No. T34935G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69804-19-9
Molecular Formula C8F13NO2
Molecular Weight (g/mol) 389.07
MDL Number MFCD30062897
InChI Key LYIPDZSLYLDLCU-UHFFFAOYNA-N
PubChem CID 112160
IUPAC Name 2,2,3,3-tetrafluoro-3-({1,1,1,2,3,3-hexafluoro-3-[(1,2,2-trifluoroethenyl)oxy]propan-2-yl}oxy)propanenitrile
SMILES FC(F)=C(F)OC(F)(F)C(F)(OC(F)(F)C(F)(F)C#N)C(F)(F)F

Specifications

Color Yellow
Boiling Point 102°C
Quantity 5 g
Formula Weight 389.07
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3-Tetrafluoro-3-[[1,1,1,2,3,3-hexafluoro-3-[(1,2,2-trifluorovinyl)oxy]propan-2-yl]oxy]propioni
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.