accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClNH2N

CAS RN 69833-17-6

IUPAC Name: hydrogen 2-(4-aminophenyl)-2-phenylacetonitrile chloride
Synonyms: hydrogen 2-(4-aminophenyl)-2-phenylacetonitrile chloride
Molecular Weight (g/mol): 244.72
Molecular Formula: C14H13ClN2
InChi Key: UMGPKQJLGUSAQX-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NC1=CC=C(C=C1)C(C#N)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results