Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Aminophenyl)phenylacetonitrile Hydrochloride 97%, Thermo Scientific™

Catalog No. AC152260050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC152260050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC152260050 Supplier Thermo Scientific Chemicals Supplier No. 152260050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69833-17-6
Molecular Formula C14H13ClN2
Molecular Weight (g/mol) 244.722
InChI Key UMGPKQJLGUSAQX-UHFFFAOYSA-N
Synonym 4-aminophenyl phenyl acetonitrile hydrochloride, 2-4-aminophenyl-2-phenylacetonitrile hydrochloride, 4-aminophenyl phenylacetonitrile hydrochloride, 4-aminophenyl phenyl acetonitrile hydrochloride 1:1, 2-p-aminophenyl-2-phenylacetonitrile monohydrochloride, 1-2-p-aminophenyl-2-phenylacetonitrile monohydrochloride
PubChem CID 116770
IUPAC Name 2-(4-aminophenyl)-2-phenylacetonitrile;hydrochloride
SMILES C1=CC=C(C=C1)C(C#N)C2=CC=C(C=C2)N.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Beige
Infrared Spectrum Authentic
Linear Formula H2NC6H4CH(C6H5)CN.HCl
Quantity 5 g
Formula Weight 244.73
Percent Purity ≥96.0%
Physical Form Fine Powder and Chunks
Chemical Name or Material (4-Aminophenyl)phenylacetonitrile hydrochloride, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.