accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNF

CAS RN 69857-37-0

IUPAC Name: 2-[4-(dicyanomethylidene)-2-fluorocyclohexa-2,5-dien-1-ylidene]propanedinitrile
Synonyms: 2-[4-(dicyanomethylidene)-2-fluorocyclohexa-2,5-dien-1-ylidene]propanedinitrile
Molecular Weight (g/mol): 222.18
Molecular Formula: C12H3FN4
InChi Key: BXPLEMMFZOKIHP-UHFFFAOYSA-N
SMILES: FC1=CC(C=CC1=C(C#N)C#N)=C(C#N)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results