Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-7,7,8,8-tetracyanoquinodimethane 98.0+%, TCI America™
SDP

Catalog No. F0509100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0509100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F0509100MG Supplier TCI America Supplier No. F0509100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69857-37-0
Molecular Formula C12H3FN4
Molecular Weight (g/mol) 222.18
MDL Number MFCD08276377
InChI Key BXPLEMMFZOKIHP-UHFFFAOYSA-N
Synonym FTCNQ
PubChem CID 19748576
IUPAC Name 2-[4-(dicyanomethylidene)-2-fluorocyclohexa-2,5-dien-1-ylidene]propanedinitrile
SMILES FC1=CC(C=CC1=C(C#N)C#N)=C(C#N)C#N

Specifications

Melting Point 218°C
Quantity 100 mg
UN Number 3439
Formula Weight 222.18
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-7,7,8,8-tetracyanoquinodimethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.