accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOON

CAS RN 699-98-9

IUPAC Name: 5H,7H-furo[3,4-b]pyridine-5,7-dione
Synonyms: furo[3,4-b]pyridine-5,7-dione
Molecular Weight (g/mol): 149.11
Molecular Formula: C7H3NO3
InChi Key: MCQOWYALZVKMAR-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=NC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)