Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3-Pyridinedicarboxylic anhydride

Catalog No. P6440525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P6440525G 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P6440525G Supplier Sigma Aldrich Supplier No. P6440525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-buten-1-ol was used as a starting reagent in asymmetric total synthesis of natural seimatopolide b
  • It was also used in the synthesis of catalytic bimetallic nanoparticles

Specifications

CAS 699-98-9
Melting Point 137°C to 139°C (lit.)
Molecular Formula C7H3NO3
Linear Formula C7H3NO3
MDL Number MFCD00005915
Quantity 25 mL
Synonym Quinolinic anhydride
Molecular Weight (g/mol) 149.1
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.