CAS RN 69912-79-4
Molecular sieves 5A, 8 to 12 mesh
CAS: 69912-79-4 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.14 MDL Number: MFCD00147627 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 SMILES: [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
Molecular sieves, 5A, 1-2mm (0.04-0.08in) dia. pellets
CAS: 69912-79-4 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.14 MDL Number: MFCD00147627 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
Molecular sieves, 5A, 3-5mm (0.12-0.20in) beads
CAS: 69912-79-4 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.14 MDL Number: MFCD00147627 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
Molecular sieves 5A, powder <50 micron
CAS: 69912-79-4 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.14 MDL Number: MFCD00147627 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
Molecular sieves 5A, 4 to 8 mesh
CAS: 69912-79-4 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.14 MDL Number: MFCD00147627 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
Molecular sieves, 5A, 3-4mm (0.12-0.16in) dia. pellets
CAS: 69912-79-4 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.14 MDL Number: MFCD00147627 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
Molecular Sieves, 5Å, Solstice
CAS: 69912-79-4 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.14 MDL Number: MFCD00147627 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 SMILES: [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
Molecular Sieves, 5Å, Pellets, 1.6mm, Solstice
CAS: 69912-79-4 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.14 MDL Number: MFCD00147627 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
Molecular Sieves, 5 Å Pellets, 3.2mm, Solstice
CAS: 69912-79-4 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.14 MDL Number: MFCD00147627 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
Molecular Sieves, 5Å, Undried, Solstice
CAS: 69912-79-4 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.14 MDL Number: MFCD00147627 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O