accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6992-39-8

IUPAC Name: 2-amino-2-(hydroxymethyl)propane-1,3-diol; phosphoric acid
Synonyms: phosphoric acid; tris buffer
Molecular Weight (g/mol): 219.13
Molecular Formula: C4H14NO7P
InChi Key: JLEXUIVKURIPFI-UHFFFAOYSA-N
SMILES: OP(O)(O)=O.NC(CO)(CO)CO
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results