Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris-phosphate, 98%

Catalog No. AC264952500
Change view
Click to view available options
Quantity:
250 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC264952500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC264952500 Supplier Thermo Scientific Chemicals Supplier No. 264952500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6992-39-8 | C4H14NO7P | 219.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6992-39-8
Molecular Formula C4H14NO7P
Molecular Weight (g/mol) 219.13
MDL Number MFCD00043285
InChI Key JLEXUIVKURIPFI-UHFFFAOYSA-N
Synonym tris phosphate, tris phosphate monobasic, tris hydroxymethyl aminomethane phosphate, 2-hydroxy-1,1-bis hydroxymethyl ethyl ammonium dihydrogen phosphate, 2-amino-2-hydroxymethyl propane-1,3-diol; phosphoric acid, mono tris hydroxymethyl aminomethane phosphate, phosphoric acid; tris buffer, trisphosphate, c4h11no3.h3o4p, mono-tris phosphate
PubChem CID 3083992
ChEBI CHEBI:67098
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;phosphoric acid
SMILES OP(O)(O)=O.NC(CO)(CO)CO

Specifications

Melting Point 168.0°C to 172.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NH2C(CH2OH)3·H3PO4
Quantity 250 g
Solubility Information Solubility in water: 1g/100 mL.
Formula Weight 219.13
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tris-phosphate, 98%
Show More Show Less
EINECSNumber 230-260-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.