accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOHHOOOH(E/Z)

CAS RN 69989-75-9

IUPAC Name: 5-[1-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)ethyl]-2-hydroxybenzoic acid
Synonyms: 5-[1-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)ethyl]-2-hydroxybenzoic acid
Molecular Weight (g/mol): 300.27
Molecular Formula: C16H12O6
InChi Key: GZUNJUHSRLTITI-UHFFFAOYSA-N
SMILES: CC(C1=CC(C(O)=O)=C(O)C=C1)=C1C=CC(=O)C(=C1)C(O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results