CAS RN 6999-03-7
IUPAC Name:
(4-bromophenyl)trimethylsilane
Synonyms:
(4-bromophenyl)trimethylsilane
Molecular Weight (g/mol):
229.19
Molecular Formula:
C9H13BrSi
InChi Key:
UKTSSJJZFVGTCG-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)C1=CC=C(Br)C=C1
1
–
1
of
1
results
1
1-Bromo-4-(trimethylsilyl)benzene 97.0+%, TCI America™
CAS: 6999-03-7 Molecular Formula: C9H13BrSi Molecular Weight (g/mol): 229.19 MDL Number: MFCD00094165 InChI Key: UKTSSJJZFVGTCG-UHFFFAOYSA-N Synonym: (4-Bromophenyl)trimethylsilane PubChem CID: 138897 IUPAC Name: (4-bromophenyl)-trimethylsilane SMILES: C[Si](C)(C)C1=CC=C(C=C1)Br