Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-4-(trimethylsilyl)benzene 97.0+%, TCI America™
SDP

Catalog No. B33575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33575G 5 g
B335725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33575G Supplier TCI America Supplier No. B33575G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6999-03-7
Molecular Formula C9H13BrSi
Molecular Weight (g/mol) 229.19
MDL Number MFCD00094165
InChI Key UKTSSJJZFVGTCG-UHFFFAOYSA-N
Synonym (4-Bromophenyl)trimethylsilane
PubChem CID 138897
IUPAC Name (4-bromophenyl)-trimethylsilane
SMILES C[Si](C)(C)C1=CC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 230°C
Quantity 5 g
Formula Weight 229.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-(trimethylsilyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.