accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClClClHOClClCl

CAS RN 70-30-4

IUPAC Name: 3,4,6-trichloro-2-[(2,3,5-trichloro-6-hydroxyphenyl)methyl]phenol
Synonyms: hexachlorophene hexachlorophenum armohex caswell no. 566 hilo flea powder Pre-OP hexophene fostril hexascrub surofene
Molecular Weight (g/mol): 406.89
Molecular Formula: C13H6Cl6O2
InChi Key: ACGUYXCXAPNIKK-UHFFFAOYSA-N
SMILES: OC1=C(CC2=C(Cl)C(Cl)=CC(Cl)=C2O)C(Cl)=C(Cl)C=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results