Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexachlorophene Solution, 5000μg/mL in methanol, Ultra Scientific

Catalog No. USEPA1129
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA-1129 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. US-EPA-1129 Supplier Agilent Technologies Supplier No. EPA11291
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 70-30-4 , 67-56-1
Molecular Formula C13H6Cl6O2
Molecular Weight (g/mol) 406.889
InChI Key ACGUYXCXAPNIKK-UHFFFAOYSA-N
PubChem CID 3598
ChEBI CHEBI:5693
IUPAC Name 3,4,6-trichloro-2-[(2,3,5-trichloro-6-hydroxyphenyl)methyl]phenol
SMILES C1=C(C(=C(C(=C1Cl)Cl)CC2=C(C(=CC(=C2Cl)Cl)Cl)O)O)Cl

Specifications

CAS Max % 99.36, 0.63
Analyte or Component Names methanol,hexachlorocyclopentadiene
Element Methanol, Hexachlorocyclopentadiene
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 5000μg/mL
Chemical Name or Material Hexachlorophene Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.