accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 700-93-6

IUPAC Name: 3-(1,3-thiazolidin-2-yl)pyridine
Synonyms: 3-(1,3-thiazolidin-2-yl)pyridine
Molecular Weight (g/mol): 166.24
Molecular Formula: C8H10N2S
InChi Key: SMYGUEYUDFIZCL-UHFFFAOYNA-N
SMILES: C1CSC(N1)C1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results