Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Pyridyl)thiazolidine, 97%, Thermo Scientific™

Catalog No. AAH62079MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62079MD 250 mg
AAH6207903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62079MD Supplier Thermo Scientific Chemicals Supplier No. H62079MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-93-6
Molecular Formula C8H10N2S
Molecular Weight (g/mol) 166.24
MDL Number MFCD01691463
InChI Key SMYGUEYUDFIZCL-UHFFFAOYNA-N
Synonym 3-1,3-thiazolidin-2-yl pyridine, 3-2-thiazolidinyl pyridine, pyridine, 3-2-thiazolidinyl, 2-3-pyridyl thiazolidine
PubChem CID 120769
IUPAC Name 2-pyridin-3-yl-1,3-thiazolidine
SMILES C1CSC(N1)C1=CC=CN=C1

Specifications

Quantity 250 mg
Formula Weight 166.24
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-(3-Pyridyl)thiazolidine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber UT8571100
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.