accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2S

CAS RN 70010-48-9

IUPAC Name: 4-(thiophen-2-yl)aniline
Synonyms: 4-(thiophen-2-yl)aniline
Molecular Weight (g/mol): 175.25
Molecular Formula: C10H9NS
InChi Key: WPNWHSQJRALLBJ-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results