accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7005-47-2

IUPAC Name: hydrogen 2-(dimethylamino)-2-methylpropan-1-ol chloride
Synonyms: dimethylamino methylpropanol
Molecular Weight (g/mol): 153.65
Molecular Formula: C6H16ClNO
InChi Key: QVNXEWQCNPSJLG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)C(C)(C)CO
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)