Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(N,N-Dimethylamino)isobutanol, MP Biomedicals

Catalog No. ICN208382 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN208382 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN208382 Supplier MP Biomedicals, Inc Supplier No. 0520838280

Chemical Identifiers

CAS 7005-47-2
Molecular Formula C6H16ClNO
Molecular Weight (g/mol) 153.65
MDL Number MFCD00021978
InChI Key QVNXEWQCNPSJLG-UHFFFAOYSA-N
Synonym dmamp, 2-dimethylamino-2-methylpropan-1-ol, 1-propanol, 2-dimethylamino-2-methyl, usaf cs-1, 2-dimethylamino-2-methyl-1-propanol, 2-dimethylamino-2-methylpropanol, unii-6zwg40uq9e, 2-n,n-dimethylamino isobutanol, 6zwg40uq9e
PubChem CID 23435
IUPAC Name hydrogen 2-(dimethylamino)-2-methylpropan-1-ol chloride
SMILES [H+].[Cl-].CN(C)C(C)(C)CO

Specifications

Quantity 100 g
Formula Weight 117.19
Chemical Name or Material 2-(N, N-Dimethylamino)Isobutanol
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.