accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70069-04-4

IUPAC Name: (1,4,7,10,13,16-hexaoxacyclooctadecan-2-yl)methanol
Synonyms: 1,4,7,10,13,16-hexaoxacyclooctadecan-2-ylmethanol
Molecular Weight (g/mol): 294.34
Molecular Formula: C13H26O7
InChi Key: HFRGASADQCZXHH-UHFFFAOYNA-N
SMILES: OCC1COCCOCCOCCOCCOCCO1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)