accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70086-62-3

IUPAC Name: 1,1,5,5-tetrafluoropentane-2,4-dione
Synonyms: 1,1,5,5-tetrafluoropentane-2,4-dione
Molecular Weight (g/mol): 172.08
Molecular Formula: C5H4F4O2
InChi Key: FTTDMWQQHZSARH-UHFFFAOYSA-N
SMILES: FC(F)C(=O)CC(=O)C(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results