Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,5,5-Tetrafluoro-2,4-pentanedione 97.0+%, TCI America™
SDP

Catalog No. T33305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T33305G Supplier TCI America Supplier No. T33305G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 70086-62-3
Molecular Formula C5H4F4O2
Molecular Weight (g/mol) 172.08
MDL Number MFCD03419795
InChI Key FTTDMWQQHZSARH-UHFFFAOYSA-N
PubChem CID 12640721
IUPAC Name 1,1,5,5-tetrafluoropentane-2,4-dione
SMILES FC(F)C(=O)CC(=O)C(F)F

Specifications

Color Yellow
Boiling Point 127°C
Quantity 5 g
UN Number 2920
Formula Weight 172.08
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,5,5-Tetrafluoro-2,4-pentanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.