accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COON

CAS RN 70136-02-6

IUPAC Name: octyl pyridine-3-carboxylate
Synonyms: nicotinicacid,octylester
Molecular Weight (g/mol): 235.33
Molecular Formula: C14H21NO2
InChi Key: TYBQVEHLLOHMLT-UHFFFAOYSA-N
SMILES: CCCCCCCCOC(=O)C1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results