Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octyl Nicotinate 98.0+%, TCI America™
SDP

Catalog No. N03305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N03305G Supplier TCI America Supplier No. N03305G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 70136-02-6
Molecular Formula C14H21NO2
Molecular Weight (g/mol) 235.327
MDL Number MFCD00059783
InChI Key TYBQVEHLLOHMLT-UHFFFAOYSA-N
Synonym Nicotinic Acid n-Octyl Ester
PubChem CID 96462
IUPAC Name octyl pyridine-3-carboxylate
SMILES CCCCCCCCOC(=O)C1=CN=CC=C1

Specifications

Color Yellow
Boiling Point 176°C
Quantity 5 g
Formula Weight 235.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material n-Octyl Nicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.