accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOCl

CAS RN 701918-26-5

IUPAC Name: N-benzyl-4-chloro-N-phenylbenzamide
Synonyms: N-benzyl-4-chloro-N-phenylbenzamide
Molecular Weight (g/mol): 321.8
Molecular Formula: C20H16ClNO
InChi Key: LDDXRNJXGXWUQJ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)N(CC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results