accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 702-62-5

IUPAC Name: 1,3-diazaspiro[4.5]decane-2,4-dione
Synonyms: 1,3-diazaspiro[4.5]decane-2,4-dione
Molecular Weight (g/mol): 168.2
Molecular Formula: C8H12N2O2
InChi Key: NERNEXMEYQFFHU-UHFFFAOYSA-N
SMILES: O=C1NC(=O)C2(CCCCC2)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results