Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diazaspiro[4.5]decane-2,4-dione, 97%, Thermo Scientific™

Catalog No. JFD02410DE
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
JFD02410DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JFD02410DE Supplier Thermo Scientific Chemicals Supplier No. JFD02410DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 702-62-5
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.196
MDL Number MFCD00022404
InChI Key NERNEXMEYQFFHU-UHFFFAOYSA-N
Synonym 1,3-diazaspiro 4.5 decane-2,4-dione, 5,5-pentamethylenehydantoin, hydantoin, pentamethylene, 5,5'-cyclohexanespirohydantoin, 5,5-pentamethylenespirohydantoin, hydantoin, 5,5-pentamethylene, cyclohexanespiro-5'-hydantoin, spiro cyclohexane-1,5'-hydantoin, 2,4-diazaspiro 4.5 decane-1,3-dione, cyclohexane-spiro-5-hydantoin
PubChem CID 12799
IUPAC Name 1,3-diazaspiro[4.5]decane-2,4-dione
SMILES C1CCC2(CC1)C(=O)NC(=O)N2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 168.19
Percent Purity 97%
Chemical Name or Material 1, 3-Diazaspiro[4.5]decane-2, 4-dione
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.