accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPOCH3Br

CAS RN 70219-09-9

IUPAC Name: [(3,4-dimethoxyphenyl)methyl]triphenylphosphanium bromide
Synonyms: [(3,4-dimethoxyphenyl)methyl]triphenylphosphanium bromide
Molecular Weight (g/mol): 493.38
Molecular Formula: C27H26BrO2P
InChi Key: BLALYSCMLUQKSG-UHFFFAOYSA-M
SMILES: [Br-].COC1=C(OC)C=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results