Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3,4-Dimethoxybenzyl)triphenylphosphonium Bromide 98.0+%, TCI America™
SDP

Catalog No. D40725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40725G Supplier TCI America Supplier No. D40725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 70219-09-9
Molecular Formula C27H26BrO2P
Molecular Weight (g/mol) 493.381
InChI Key BLALYSCMLUQKSG-UHFFFAOYSA-M
PubChem CID 11049284
IUPAC Name (3,4-dimethoxyphenyl)methyl-triphenylphosphanium;bromide
SMILES COC1=C(C=C(C=C1)C[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)OC.[Br-]

Specifications

Melting Point 241°C
Color White-Yellow
Quantity 5 g
Formula Weight 493.38
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (3,4-Dimethoxybenzyl)triphenylphosphonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.