accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSO

CAS RN 7022-45-9

IUPAC Name: 2-(methylsulfanyl)benzaldehyde
Synonyms: 2-(methylsulfanyl)benzaldehyde
Molecular Weight (g/mol): 152.21
Molecular Formula: C8H8OS
InChi Key: XIOBUABQJIVPCQ-UHFFFAOYSA-N
SMILES: CSC1=CC=CC=C1C=O
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results