Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylthio)benzaldehyde 97.0+%, TCI America™
SDP

Catalog No. M20941G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M20941G Supplier TCI America Supplier No. M20941G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7022-45-9
Molecular Formula C8H8OS
Molecular Weight (g/mol) 152.211
MDL Number MFCD00196822
InChI Key XIOBUABQJIVPCQ-UHFFFAOYSA-N
PubChem CID 286443
IUPAC Name 2-methylsulfanylbenzaldehyde
SMILES CSC1=CC=CC=C1C=O

Specifications

Color Yellow
Boiling Point 149°C
Quantity 1 g
Formula Weight 152.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Methylthio)benzaldehyde
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.