accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 702680-17-9

IUPAC Name: 2-methyl-2-[3-(1-{2-[4-(propan-2-yl)phenyl]acetyl}piperidin-3-yl)phenoxy]propanoic acid
Synonyms: 2-(3-{1-[2-(4-isopropylphenyl)acetyl]piperidin-3-yl}phenoxy)-2-methylpropanoic acid
Molecular Weight (g/mol): 423.55
Molecular Formula: C26H33NO4
InChi Key: OISHBINQIFNIPV-UHFFFAOYNA-N
SMILES: CC(C)C1=CC=C(CC(=O)N2CCCC(C2)C2=CC(OC(C)(C)C(O)=O)=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results