Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CP 775146
SDP

Catalog No. 419010 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-901-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 41-901-0 Supplier Tocris Bioscience™ Supplier No. 4190/10

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective, high affinity PPARα agonist

Selective, high affinity PPARα agonist (Ki values are 24.5 nM for PPARα and >10 μM for PPARβ and PPARγ in vitro). Exhibits hypolipidemic activity in vivo.

Chemical Identifiers

CAS 702680-17-9
Molecular Formula C26H33NO4
Molecular Weight (g/mol) 423.553
InChI Key OISHBINQIFNIPV-JOCHJYFZSA-N
Synonym 2-methyl-2-3-3s-1-2-4-1-methylethyl phenyl acetyl-3-piperidinyl phenoxy-propanoic acid, 2-3-3s-1-2-4-isopropylphenyl acetyl piperidin-3-yl phenoxy-2-methylpropanoic acid, 2-methyl-2-3-3s-1-4-propan-2-yl phenyl acetyl piperidin-3-yl phenoxy propanoic acid, s-2-3-1-2-4-isopropylphenyl acetyl piperidin-3-yl phenoxy-2-methylpropanoic acid, d01kpn, 3r,4r-1-cyanoacetyl-4-methyl-n-methyl-n-1h-pyrrolo 2,3-d pyrimidin-4-yl-3-piperidinamine 2-hydroxy-1,2,3-propanetricarboxylate, 3r,4r-4-methyl-3-methyl-7h-pyrrolo 2,3-d pyrimidin-4-ylamino-, a-oxo-1-piperidinepropanenitrile, 2-methyl-2-3-3s-1-2-4-propan-2-ylphenyl acetyl piperidin-3-yl phenoxy propanoic acid
PubChem CID 10410059
IUPAC Name 2-methyl-2-[3-[(3S)-1-[2-(4-propan-2-ylphenyl)acetyl]piperidin-3-yl]phenoxy]propanoic acid
SMILES CC(C)C1=CC=C(C=C1)CC(=O)N2CCCC(C2)C3=CC(=CC=C3)OC(C)(C)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 423.54
Percent Purity >99%
Chemical Name or Material CP 775146
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.