accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClN

CAS RN 703-61-7

IUPAC Name: 2,4-dichloroquinoline
Synonyms: 2,4-dichloroquinoline
Molecular Weight (g/mol): 198.05
Molecular Formula: C9H5Cl2N
InChi Key: QNBJYUUUYZVIJP-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C2C=CC=CC2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results