accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOHP

CAS RN 70340-04-4

IUPAC Name: [(2-hydroxyphenyl)methyl]triphenylphosphanium bromide
Synonyms: [(2-hydroxyphenyl)methyl]triphenylphosphanium bromide
Molecular Weight (g/mol): 449.33
Molecular Formula: C25H22BrOP
InChi Key: FKKZGQXMWVGPMH-UHFFFAOYSA-N
SMILES: [Br-].OC1=CC=CC=C1C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results