Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Hydroxybenzyl)triphenylphosphonium Bromide 97.0+%, TCI America™
SDP

Catalog No. H12405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12405G Supplier TCI America Supplier No. H12405G
Add to Cart
Add to Cart

Specifications

CAS 70340-04-4
Melting Point 243°C
Molecular Formula C25H22BrOP
Quantity 5 g
MDL Number MFCD00011902
Synonym 2-hydroxybenzyl triphenylphosphonium bromide, 2-hydroxyphenyl methyl-triphenylphosphanium bromide, 2-hydroxyphenyl methyl triphenylphosphanium bromide, phosphonium, 2-hydroxyphenyl methyl triphenyl-, bromide, acmc-209oe4, 2-hydroxybenzyltriphenylphosphonium bromide, 2-hydroxybenzyl triphenylphosphoniumbromide, 2-hydroxybenzyl triphenyl phosphonium bromide, ortho hydroxybenzyltriphenyl-phosphonium bromide, 2-hydroxyphenyl methyl-triphenylphosphonium bromide
InChI Key FKKZGQXMWVGPMH-UHFFFAOYSA-N
SMILES [Br-].OC1=CC=CC=C1C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name [(2-hydroxyphenyl)methyl]triphenylphosphanium bromide
Molecular Weight (g/mol) 449.33
PubChem CID 2724142
Formula Weight 449.33
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Hydroxybenzyl)triphenylphosphonium Bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.