accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70445-33-9

IUPAC Name: 3-[(2-ethylhexyl)oxy]propane-1,2-diol
Synonyms: ethylhexylglycerin
Molecular Weight (g/mol): 204.31
Molecular Formula: C11H24O3
InChi Key: NCZPCONIKBICGS-UHFFFAOYNA-N
SMILES: CCCCC(CC)COCC(O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results