Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Ethylhexyloxy)-1,2-propanediol 98.0+%, TCI America™
SDP

Catalog No. E104425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E104425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E104425ML Supplier TCI America Supplier No. E104425ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 70445-33-9
Molecular Formula C11H24O3
Molecular Weight (g/mol) 204.31
MDL Number MFCD03790878
InChI Key NCZPCONIKBICGS-UHFFFAOYNA-N
Synonym Glycerol alpha-(2-Ethylhexyl) Ether
PubChem CID 9859093
IUPAC Name 3-[(2-ethylhexyl)oxy]propane-1,2-diol
SMILES CCCCC(CC)COCC(O)CO

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 204.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(2-Ethylhexyloxy)-1,2-propanediol
EINECSNumber (2)-0414
RTECSNumber TY6462200
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.