accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHClClHONHHNHNNHOHOOHHOO

CAS RN 70476-82-3

IUPAC Name: dihydrogen 1,4-dihydroxy-5,8-bis({2-[(2-hydroxyethyl)amino]ethyl}amino)-9,10-dihydroanthracene-9,10-dione dichloride
Synonyms: mitoxantrone
Molecular Weight (g/mol): 517.4
Molecular Formula: C22H30Cl2N4O6
InChi Key: ZAHQPTJLOCWVPG-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].OCCNCCNC1=CC=C(NCCNCCO)C2=C1C(=O)C1=C(O)C=CC(O)=C1C2=O
Product Line 
  • (1)
Product Type 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results