accessibility menu, dialog, popup

UserName

Viewing profile as user:
Br

CAS RN 7051-34-5

IUPAC Name: (bromomethyl)cyclopropane
Synonyms: (bromomethyl)cyclopropane
Molecular Weight (g/mol): 135
Molecular Formula: C4H7Br
InChi Key: AEILLAXRDHDKDY-UHFFFAOYSA-N
SMILES: BrCC1CC1
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (3)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (6)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)