Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Bromomethyl)cyclopropane

Catalog No. 2424035G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-240-35G 5 g
24-240-31G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-240-35G Supplier Sigma Aldrich Supplier No. 2424035G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The coupling reaction of (bromomethyl)cyclopropane with phenylmagnesium bromide was studied.(bromomethyl)cyclopropane was used as a synthetic building block for the introduction of the cyclopropylmethyl group
  • It was also used in the synthesis of 1,4-dienes via iron-catalyzed cross-coupling with alkenyl grignard reagents

Specifications

CAS 7051-34-5
Density 1.392 g/mL (at 25°C (literature))
Boiling Point 105°C to 107°C (lit.)
Molecular Formula C4H7Br
Linear Formula C3H5CH2Br
Refractive Index n20/D 1.457 (literature)
MDL Number MFCD00001306
Quantity 5 g
Synonym Cyclopropylmethyl bromide
Molecular Weight (g/mol) 135
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.