accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70523-22-7

IUPAC Name: (2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)boronic acid
Synonyms: 2,4-dioxo-1,3-dihydropyrimidin-5-ylboronic acid
Molecular Weight (g/mol): 155.9
Molecular Formula: C4H5BN2O4
InChi Key: PVEJOCQTIVCDNO-UHFFFAOYSA-N
SMILES: OB(O)C1=CNC(=O)NC1=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results